Learn More
Medchemexpress LLC DNA2 inhibitor C5 | 35973-25-2 | 98.5% | 100 MG
SDP
Catalog No. 5000342063
Encompass_Preferred
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. It can be used in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Suitable for breast cancer and colorectal cancer research
  • Displays strong synergistic effect in inhibiting MCF7 cell survival
  • Shows synthetic lethality with deficiencies in FEN1, EXO1, or MRE11 genes in mouse model
Catalog No. 50-003-42063 Supplier Medchemexpress LLC Supplier No. HY128729100MG
DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. It can be used in breast cancer and colorectal cancer research.
  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2
  • Suitable for breast cancer and colorectal cancer research
  • Displays strong synergistic effect in inhibiting MCF7 cell survival
  • Shows synthetic lethality with deficiencies in FEN1, EXO1, or MRE11 genes in mouse model