Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DNA2 inhibitor C5 | 35973-25-2 | 98.5% | 50 MG
SDP

Catalog No. 5000341343
Encompass_Preferred

DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. It can be used in breast cancer and colorectal cancer research.

  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits DNA2 nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities
  • IC50 of 20 μM against DNA2 nuclease
  • Can be used in breast cancer and colorectal cancer research
  • Shows synergistic effect in inhibiting MCF7 cell survival
  • High purity of 98.51%
  • Molecular weight: 234.17
  • Chemical formula: C10H6N2O5
  • Appearance: Solid, light yellow to yellow

Catalog No. 50-003-41343 Supplier Medchemexpress LLC Supplier No. HY12872950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA2 inhibitor C5 is a potent, competitive, and specific DNA2 nuclease inhibitor with an IC50 of 20 μM. It inhibits the nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities of DNA2. It can be used in breast cancer and colorectal cancer research.

  • Potent, competitive, and specific DNA2 nuclease inhibitor
  • Inhibits DNA2 nuclease, DNA-dependent ATPase, helicase, and DNA-binding activities
  • IC50 of 20 μM against DNA2 nuclease
  • Can be used in breast cancer and colorectal cancer research
  • Shows synergistic effect in inhibiting MCF7 cell survival
  • High purity of 98.51%
  • Molecular weight: 234.17
  • Chemical formula: C10H6N2O5
  • Appearance: Solid, light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.