Learn More
TARGETMOL CHEMICALS INC DNA2 inhibitor C5 5MG
SDP
Catalog No. 502878267
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DNA2 inhibitor C5 is a promising lead compound to develop sensitizers for cancer chemotherapeutics that cause replication stress and it also is a competitive and specific inhibitor of DNA2 nuclease activity (IC50 20 uM). DNA2 inhibitor C5 inhibits nuclease, DNA dependent ATPase, helicase, and DNA binding activities of DNA2. Purity 98%
Catalog No. 50-287-8267 Supplier TARGETMOL CHEMICALS INC Supplier No. T151465MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DNA2 inhibitor C5 is a promising lead compound to develop sensitizers for cancer chemotherapeutics that cause replication stress and it also is a competitive and specific inhibitor of DNA2 nuclease activity (IC50 20 uM). DNA2 inhibitor C5 inhibits nuclease, DNA dependent ATPase, helicase, and DNA binding activities of DNA2. Purity 98%