Learn More
Medchemexpress LLC HY-128729 5mg Medchemexpress, DNA2 inhibitor C5 CAS:35973-25-2 Purity:>98%
SDP
Catalog No. 501969766
Encompass_Preferred
Medchemexpress, HY-128729 5mg DNA2 inhibitor C5 CAS:35973-25-2 DNA2 inhibitor C5 is a potent, competitive and specific inhibitor of DNA2 nuclease activity with an IC50 of 20 μM. DNA2 inhibitor C5 inhibits nuclease, DNA dependent ATPase, helicase, and DNA binding activities of DNA2. DNA2 inhibitor C5 is a promising lead compound to develop sensitizers for cancer chemotherapeutics that cause replication stress. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9766 Supplier Medchemexpress LLC Supplier No. HY1287295MG
Add to Cart
Add to Cart
Medchemexpress, HY-128729 5mg DNA2 inhibitor C5 CAS:35973-25-2 DNA2 inhibitor C5 is a potent, competitive and specific inhibitor of DNA2 nuclease activity with an IC50 of 20 μM. DNA2 inhibitor C5 inhibits nuclease, DNA dependent ATPase, helicase, and DNA binding activities of DNA2. DNA2 inhibitor C5 is a promising lead compound to develop sensitizers for cancer chemotherapeutics that cause replication stress. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.