Learn More
Aviva Systems Biology Corp DNA2L Antibody - middle region (ARP36549_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36549_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNA2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DNA2L. Alternate names: DNA2L, FLJ10063, KIAA0083, MGC133297ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56184