Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle Biosciences Inc DNAbsolute 5mL, 5 mL Kit
SDP

Catalog No. 5000053070
Encompass

DNAbsolute - column-free DNA extraction buffer (5mL)

Catalog No. 50-000-53070 Supplier Eagle Biosciences Inc Supplier No. ABS5
May include imposed supplier surcharges.
Add to Cart
Add to Cart

DNAbsolute - column-free DNA extraction buffer (5mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.