Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, DNACore AAV-Q LC Analytical Column, 10um, 4.6 x 50 mm, PEEK
Catalog No. NC3587981
Encompass_Preferred
DNACore AAV-Q is a family of high performance, strong anion exchange chromatography columns, designed for empty/full ratio analysis of capsid content in different AAV serotypes. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC3587981 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. D301100AAV04605P
DNACore AAV-Q is a family of high performance, strong anion exchange chromatography columns, designed for empty/full ratio analysis of capsid content in different AAV serotypes. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.