Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, DNACore AAV-SEC LC Analytical Column, 3um, 4.6 x 300 mm, Stainless Steel
Catalog No. NC3587980
Encompass_Preferred
DNACore AAV-SEC is a family of high performance, size exclusion chromatography columns, designed for aggregate analysis of AAVs in different serotypes. The column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, wide-pore, monodispersed silica particles, combined with well-established column packing processes.
Catalog No. NC3587980 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. D203030AAV04630S
DNACore AAV-SEC is a family of high performance, size exclusion chromatography columns, designed for aggregate analysis of AAVs in different serotypes. The column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, wide-pore, monodispersed silica particles, combined with well-established column packing processes.