Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, DNACore NP-Q LC Analytical Column, 5um, 4.6 x 100 mm, PEEK
Catalog No. NC3587986
Encompass_Preferred
DNACore NP-Q is a family of high-performance, strong anion-exchange columns designed for analysis of oligonucleotides, DNAs, mRNAs and plasmid topology. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.
Catalog No. NC3587986 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. D30105000004610P
DNACore NP-Q is a family of high-performance, strong anion-exchange columns designed for analysis of oligonucleotides, DNAs, mRNAs and plasmid topology. Its column technology involves the creation of a hydrophilic quaternary ammonium functional layer on the surface of monodispersed, nonporous, spherical PS/DVB particles, combined with well-established column packing processes.