Learn More
Beckman Coulter Beckman Coulter DNAdvance Kit - Genomic DNA extraction from tissue, 9,600 preps (A48706)
Catalog No. NC1978063
Encompass
Beckman Coulter DNAdvance Kit (A48706) is a high-throughput genomic DNA extraction reagent kit that uses SPRI paramagnetic bead chemistry to isolate high-quality DNA from mammalian tissue samples. It is designed for automation on liquid-handling workstations and produces DNA compatible with PCR, qPCR, SNP genotyping, and next-generation sequencing.
  • High-throughput capacity: 9,600 preps per kit.
  • Technology: SPRI paramagnetic bead-based DNA isolation.
  • Sample types: mammalian tissue including brain, kidney, liver, lung, and tail.
  • Throughput: processes three 96-well plates in ~1 hour using automation; 15 minutes hands-on time for 96 samples; 1 hour 46 minutes total run time for a 96-well plate.
  • Typical yield: isolates up to 40 μg DNA per prep.
  • Compatible with downstream analyses: PCR, qPCR, SNP genotyping, sequencing.
Catalog No. NC1978063 Supplier Beckman Coulter Supplier No. A48706
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Beckman Coulter DNAdvance Kit (A48706) is a high-throughput genomic DNA extraction reagent kit that uses SPRI paramagnetic bead chemistry to isolate high-quality DNA from mammalian tissue samples. It is designed for automation on liquid-handling workstations and produces DNA compatible with PCR, qPCR, SNP genotyping, and next-generation sequencing.
  • High-throughput capacity: 9,600 preps per kit.
  • Technology: SPRI paramagnetic bead-based DNA isolation.
  • Sample types: mammalian tissue including brain, kidney, liver, lung, and tail.
  • Throughput: processes three 96-well plates in ~1 hour using automation; 15 minutes hands-on time for 96 samples; 1 hour 46 minutes total run time for a 96-well plate.
  • Typical yield: isolates up to 40 μg DNA per prep.
  • Compatible with downstream analyses: PCR, qPCR, SNP genotyping, sequencing.