Learn More
Biomatik Corporation Dnah5 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11047
Encompass_Preferred
Dnah5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to DNAH5. The Dnah5 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dynein Heavy Chain 5, Axonemal Protein (2364-2560Aa). Applications: ELISA, IHC, IF. Target Synonyms: DNAH5. Accession Number: Q8TE73. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9254