Learn More
Aviva Systems Biology Corp DNAH9 Antibody - C-terminal region (ARP65745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAH9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human DNAH9. Alternate names: DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20, KIAA0357ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-558