Learn More
Enzo Life Sciences DnaJ (E. coli), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ALX201144C025
Encompass_Preferred
Alternative name: Heat shock protein 40, HSP40, Heat shock protein J. Concentration: 4.3mg/ml. Purity: ≥99%. Formulation: Liquid. In 30mM HEPES/KOH, pH 8.2, containing 120mM KCl, 15mM NaCl and 4% glycerol. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-200-7938