Learn More
Enzo Life Sciences DnaJ (E. coli), (recombinant) (His-tag) (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPP640J
Encompass_Preferred
Alternative name: DNAJB2, DnaJ HSJ1, HSPF3, HSP40. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~43kDa. Formulation: Liquid. In 25mM HEPES, pH 7.5, containing 200mM potassium chloride, 10% glycerol, and 5mM DTT. Source: Produced in E. coli. E. coli DnaJ is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB Protein refolding assay.
Catalog No. 50-200-7616