Learn More
Aviva Systems Biology Corp Dnaja1 Antibody - N-terminal region (ARP54580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnaja1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Hsj2, Nedd7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-944