Learn More
Aviva Systems Biology Corp DNAJA2 Antibody - N-terminal region (ARP52128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DNAJA2. Alternate names: CPR3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2, DJ3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-642