Learn More
Aviva Systems Biology Corp DNAJA3 Antibody - C-terminal region (ARP64449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJA3. It was validated on Western Blot. . Alternate names: FLJ45758, TID1, hTid-1ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-264