Learn More
Sino Biological Recombinant Human PI3K (p110 alpha/p85 alpha) Protein (p110 alpha (E545K)/ p85 alpha, GST Tag), Active
Catalog No. 502573792
Encompass_Preferred
Recombinant full length human p110alpha (E545K) mutant subunit and full length human p85alpha wild-type subunit were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
Catalog No. 50-257-3792 Supplier Sino Biological Supplier No. P27152G500
Recombinant full length human p110alpha (E545K) mutant subunit and full length human p85alpha wild-type subunit were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.