Learn More
Aviva Systems Biology Corp DNAJB11 Antibody - C-terminal region (ARP47245_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47245_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJB11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DNAJB11. Alternate names: ABBP-2, ABBP2, EDJ, ERdj3, ERj3, HEDJ, PRO1080, UNQ537, hDj9, DJ9, Dj-9, ERj3p, hDj-9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59496