Learn More
Aviva Systems Biology Corp Dnajb12 Antibody - N-terminal region (ARP37277_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37277_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnajb12. It was validated on Western Blot. Dnajb12,DnaJ (Hsp40) homolog, subfamily B, member 12,Dj10; mDj10; Dnajb12,dnaJ homolog subfamily B member 12,dnaJ homolog subfamily B member 12,DnaJ homolog 10,OTTMUSP00000046642,OTTMUSP00000046643,OTTMUSP00000046645The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Dj10, mDj10ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-610