Learn More
Aviva Systems Biology Corp DNAJB7 Antibody - C-terminal region (ARP52805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJB7. It was validated on Western Blot. . Alternate names: DJ5, HSC3, MGC138340ApplicationWB. Species reactivity: Human
Catalog No. 50-285-769