Learn More
Aviva Systems Biology Corp DNAJB9 Antibody - N-terminal region (ARP46885_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46885_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJB9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DNAJB9. Alternate names: DKFZP564F1862, ERdj4, MDG1, MST049, MSTP049, MDG-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59428