Learn More
Aviva Systems Biology Corp DNAJC1 Antibody - middle region (ARP49711_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49711_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DNAJC1. Alternate names: DNAJL1, ERdj1, HTJ1, MGC131954, MTJ1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60154