Learn More
Aviva Systems Biology Corp Dnajc11 Antibody - C-terminal region (ARP62536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnajc11. It was validated on Western Blot. . Alternate names: E030019A03RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-706