Learn More
Aviva Systems Biology Corp DNAJC11 Antibody - C-terminal region (ARP62537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC11. It was validated on Western Blot. . Alternate names: FLJ10737, dJ126A5.1, RP1-126A5.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-707