Learn More
Aviva Systems Biology Corp DNAJC17 Antibody - N-terminal region (ARP37472_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37472_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse DNAJC17. Alternate names: C87112, D9Bwg1371e, 1700025B16RikApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56374