Learn More
Aviva Systems Biology Corp DNAJC19 Antibody - C-terminal region (ARP59815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC19. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DNAJC19. Alternate names: TIM14, TIMM14, PAM18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-839