Learn More
Aviva Systems Biology Corp DNAJC2 Antibody - C-terminal region (ARP59866_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59866_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DNAJC2. Alternate names: MPHOSPH11, MPP11, ZRF1, ZUO1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-858