Learn More
Aviva Systems Biology Corp Dnajc21 Antibody - middle region (ARP62801_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnajc21. It was validated on Western Blot. . Alternate names: 4930461P20Rik, 9930116P15RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-772