Learn More
Aviva Systems Biology Corp Dnajc25 Antibody - middle region (ARP44698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnajc25. It was validated on Western Blot. . Alternate names: 2010109C08Rik, 2010203O07Rik, RP23-211P15.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-158