Learn More
Aviva Systems Biology Corp Dnajc27 Antibody - N-terminal region (ARP56937_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56937_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnajc27. It was validated on Western Blot. . Alternate names: AI639580, C330021A05Rik, Rabj, RbjApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-238