Learn More
Aviva Systems Biology Corp DNAJC7 Antibody - C-terminal region (ARP65551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human DNAJC7. Alternate names: DJ11, DJC7, TPR2, TTC2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-516