Learn More
Aviva Systems Biology Corp DNAJC9 Antibody - C-terminal region (ARP62539_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62539_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJC9. It was validated on Western Blot. . Alternate names: HDJC9, JDD1, KIAA0974, SB73ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-708