Learn More
Enzo Life Sciences DnaK (E. coli), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP630D
Encompass_Preferred
Alternative name: Heat shock protein 70, HSP70. MW: ~70kDa. Purity: ≥95% (SDS-PAGE; Western blot). Formulation: Liquid. In 40mM TRIS, pH 7.5, containing 80mM sodium chloride, 0.8mM DTT, 0.08mM PMSF, and 20% glycerol. Source: Produced in E. coli. UniProt: P0A6Y8 (strain K12). Application: ELISA, WB, in vitro Assay, Activity assay. Long Term Storage: -80°C
Catalog No. 50-667-731