Learn More
Enzo Life Sciences DnaK (E. coli) (1-638), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX201217C100
Encompass_Preferred
Alternative name: Heat shock protein 70, HSP70. Concentration: 1mg/ml. Purity: ≥95% (SDS-PAGE). MW: ~69kDa. Formulation: Liquid. In 20mM TRIS buffer, pH 7.5, containing 100mM sodium chloride, 5mM DTT and 10% glycerol. Source: Recombinant full length DnaK produced in E. coli. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7949