Learn More
Enzo Life Sciences DnaK (E. coli) monoclonal antibody (8E2/2) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6800200
Encompass_Preferred
Clone: 8E2/2. Isotype: IgG1. Immunogen: E. coli DnaK. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: E. coli. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3211