Learn More
Enzo Life Sciences DnaK (E. coli), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ALX201143C025
Encompass_Preferred
Alternative name: Heat shock protein 70, HSP70. Purity: ≥99%. Formulation: Liquid. In 20mM TRIS/HCl, pH 7.2, containing 100mM NaCl, 40mM KCl, 0.1mM EDTA, 1mM DTT and 5% glycerol. Source: Produced in E. coli. Long Term Storage: -80°C. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles. Applications: WB .
Catalog No. 50-200-7937