Learn More
Enzo Life Sciences DnaK (E. coli) monoclonal antibody (8E2/2) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA880D
Encompass_Preferred
Clone: 8E2/2. Host: Mouse. Isotype: IgG1. Immunogen: E. coli DnaK. UniProt: P0A6Y8 (strain K12). Source: Purified from ascites. Species Reactivity: E. coli. Applications: WB. Recommended dilutions: Western Blot (1:10,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-656