Learn More
Aviva Systems Biology Corp Dnalc4 Antibody - middle region (ARP52101_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52101_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dnalc4. It was validated on Western Blot. . Alternate names: D15Ertd424e, Dnal4, Dnalc4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-635