Learn More
Aviva Systems Biology Corp DNALI1 Antibody - N-terminal region (ARP53611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DNALI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DNALI1. Alternate names: P28, dJ423B22.5, hp28ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61068