Learn More
ACROBiosystems Human CD226 Protein, Fc Tag, Lyophilized, 1 mg
Catalog No. 501658895
Encompass_Preferred
Human DNAM-1, also known as CD226 or PTA1, is a single-pass type I membrane protein containing two Ig-like C2-type domains. Expressed on the surface of natural killer cells, platelets, monocytes, and T cells, this glycoprotein mediates cellular adhesion to ligands such as CD112 and CD155 and is involved in cellular activation. This protein is expressed from HEK293 cells and contains amino acids Glu 19 to Asn 247 with a human IgG1 Fc tag at the C-terminus.
  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS
  • Endotoxin level less than 1.0 EU per μg
  • Calculated molecular weight of 52.6 kDa
  • Lyophilized in a buffer of Tris, Glycine, Arginine, and NaCl, pH 7.5 with trehalose as a protectant
  • Validated by ELISA, BLI, and FACS
Catalog No. 50-165-8895 Supplier ACROBiosystems Supplier No. DN1H52571MG
Human DNAM-1, also known as CD226 or PTA1, is a single-pass type I membrane protein containing two Ig-like C2-type domains. Expressed on the surface of natural killer cells, platelets, monocytes, and T cells, this glycoprotein mediates cellular adhesion to ligands such as CD112 and CD155 and is involved in cellular activation. This protein is expressed from HEK293 cells and contains amino acids Glu 19 to Asn 247 with a human IgG1 Fc tag at the C-terminus.
  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS
  • Endotoxin level less than 1.0 EU per μg
  • Calculated molecular weight of 52.6 kDa
  • Lyophilized in a buffer of Tris, Glycine, Arginine, and NaCl, pH 7.5 with trehalose as a protectant
  • Validated by ELISA, BLI, and FACS