Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Protein, Cynomolgus DNAM-1, His Tag, 100 μg

Catalog No. 501657605
Encompass_Preferred

Cynomolgus DNAM-1 (CD226) is a single-pass type I membrane protein containing two Ig-like C2-type domains. Expressed in human 293 cells, this recombinant protein includes a polyhistidine tag at the C-terminus. It serves as a mediator of cellular adhesion to ligands such as CD112 and CD155 and is involved in cellular activation. The protein is supplied as a lyophilized powder.

  • Purity: >90% as determined by SDS-PAGE
  • Calculated molecular weight: 28.5 kDa (migrates as 40–55 kDa under reducing conditions)
  • Endotoxin level: <1.0 EU per μg by the LAL method
  • Formulation: Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Stable for 12 months in lyophilized state at -20°C to -70°C
  • Validated for binding interactions with CD155 via ELISA and BLI

Catalog No. 50-165-7605 Supplier ACROBiosystems Supplier No. DN1C52H9100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cynomolgus DNAM-1 (CD226) is a single-pass type I membrane protein containing two Ig-like C2-type domains. Expressed in human 293 cells, this recombinant protein includes a polyhistidine tag at the C-terminus. It serves as a mediator of cellular adhesion to ligands such as CD112 and CD155 and is involved in cellular activation. The protein is supplied as a lyophilized powder.

  • Purity: >90% as determined by SDS-PAGE
  • Calculated molecular weight: 28.5 kDa (migrates as 40–55 kDa under reducing conditions)
  • Endotoxin level: <1.0 EU per μg by the LAL method
  • Formulation: Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Stable for 12 months in lyophilized state at -20°C to -70°C
  • Validated for binding interactions with CD155 via ELISA and BLI

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.