Learn More
Cayman Chemical DNARNA OxIdatIve DamageClo 1ug
SDP
Catalog No. 502942492
Encompass_Preferred
A KAT8 inhibitor selective for KAT8 over KAT2A KAT2B KAT3B KAT6A and KAT7 at 200 UM reduces H4K16Ac in HT-29 cells at 50 UM reduces the viability of HCT116 H1299 A549 and U937 cancer cells (IC50s 39.4 52.1 41 and 30.1 UM respectively)
Catalog No. 50-294-2492 Supplier Cayman Chemical Supplier No. 4011341 UG
A KAT8 inhibitor selective for KAT8 over KAT2A KAT2B KAT3B KAT6A and KAT7 at 200 UM reduces H4K16Ac in HT-29 cells at 50 UM reduces the viability of HCT116 H1299 A549 and U937 cancer cells (IC50s 39.4 52.1 41 and 30.1 UM respectively)