Learn More
Cayman Chemical DNA/RNA Oxidative Damage (High Sensitivity) ELISA Kit, 480 Well
SDP
Catalog No. 502894106
Encompass_Preferred
Oxidative damage to DNA and RNA occurs during aging and in many disease states, releasing oxidized guanine species that can be measured in urine. Cayman’s DNA/RNA Oxidative Damage ELISA detects 8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanosine, and 8-hydroxyguanine, providing a broader assessment of oxidative nucleic acid damage than assays limited to 8-hydroxy-2'-deoxyguanosine alone. Because the assay detects all three species, results should not be directly compared with LC/MS measurements of 8-hydroxy-2'-deoxyguanosine alone, as ELISA values will generally be higher.
Catalog No. 50-289-4106 Supplier Cayman Chemical Supplier No. 589320480 WELL
Oxidative damage to DNA and RNA occurs during aging and in many disease states, releasing oxidized guanine species that can be measured in urine. Cayman’s DNA/RNA Oxidative Damage ELISA detects 8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanosine, and 8-hydroxyguanine, providing a broader assessment of oxidative nucleic acid damage than assays limited to 8-hydroxy-2'-deoxyguanosine alone. Because the assay detects all three species, results should not be directly compared with LC/MS measurements of 8-hydroxy-2'-deoxyguanosine alone, as ELISA values will generally be higher.