Learn More
Cayman Chemical DNARNA OxIdatIve DamageHIg 480
SDP
Catalog No. 502892299
Encompass_Preferred
A DPP-4 inhibitor (IC50 5 nM for the human enzyme) reduces plasma DPP-4 activity and glucose levels and increases plasma insulin levels in an OGTT in wild-type and fa/fa Zucker fatty rats at 1 mg/kg reduces total ulcer length in a rat model of indomethacin-induced intestinal ulcers
Catalog No. 50-289-2299 Supplier Cayman Chemical Supplier No. 589320480 SOLID
A DPP-4 inhibitor (IC50 5 nM for the human enzyme) reduces plasma DPP-4 activity and glucose levels and increases plasma insulin levels in an OGTT in wild-type and fa/fa Zucker fatty rats at 1 mg/kg reduces total ulcer length in a rat model of indomethacin-induced intestinal ulcers