Learn More
Worthington Biochemical Corporation DNASE 1 100MG
SDP
Catalog No. NC9199796
Encompass
Deoxyribonuclease I, Chromatographically purified, Equivalent to 1X crystallized Dnase, A lyophilized powder with glycine as a stabilizer, Store at 2 to 8degreeC, Pack Size:100mg, Source: Bovine Pancreas, min. activity: 2000U/mgKunitz dry w
Catalog No. NC9199796 Supplier Worthington Biochemical Corporation Supplier No. LS002007
Deoxyribonuclease I, Chromatographically purified, Equivalent to 1X crystallized Dnase, A lyophilized powder with glycine as a stabilizer, Store at 2 to 8degreeC, Pack Size:100mg, Source: Bovine Pancreas, min. activity: 2000U/mgKunitz dry w