Learn More
Worthington Biochemical Corporation DNASE 1 100MG
SDP
Catalog No. NC9709009
Encompass
Deoxyribonuclease I, 100 MG, Purified precrystalline DNase. A lyophilized powder. Store at 2-8degreeC., Source: Bovine Pancreas, Minimum Activity: =2,000 Kunitz units per mg dry weight
Catalog No. NC9709009 Supplier Worthington Biochemical Corporation Supplier No. LS002139
Deoxyribonuclease I, 100 MG, Purified precrystalline DNase. A lyophilized powder. Store at 2-8degreeC., Source: Bovine Pancreas, Minimum Activity: =2,000 Kunitz units per mg dry weight