Learn More
Sigma Aldrich Fine Chemicals Biosciences DNase I, 100 mg
Catalog No. NC1839861
Encompass
During tissue dissociation, parts of the cells are lysed resulting in a release of DNA. Monomolecular DNA may cause clumping of cells. Addition of DNase I to the dissociation buffer leads to a degradation of this extracellular DNA, therby avoiding the loss of cells from undesired clumping.
Catalog No. NC1839861 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11284932001
During tissue dissociation, parts of the cells are lysed resulting in a release of DNA. Monomolecular DNA may cause clumping of cells. Addition of DNase I to the dissociation buffer leads to a degradation of this extracellular DNA, therby avoiding the loss of cells from undesired clumping.