Learn More
STEMCELL Technologies DNase I, ACF, Size: 100000 U
SDP
Supplier:  STEMCELL Technologies 07474
Encompass
Deoxyribonuclease I (DNase I), Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials and purified by chromatography. DNase I is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds. DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage. DNase I, ACF preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5’-phosphate and 3’-hydroxyl groups (Bernardi et al.). DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium l (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.).
Catalog No. NC1314982
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov