Learn More
Macherey-Nagel rDNase Set
Catalog No. NC9605890
Encompass_Preferred
rDNase Set Recombinant DNase and rDNase Reaction Buffer, For RNA minipreparations with NucleoSpin kits
Catalog No. NC9605890 Supplier Macherey-Nagel Supplier No. 740963
rDNase Set Recombinant DNase and rDNase Reaction Buffer, For RNA minipreparations with NucleoSpin kits